7-Acetoxy-4-methylcoumarin ≥99% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 4-Methyl-2-oxo-2H-chromen-7-yl acetate, 4-Methylumbelliferyl acetate

CDX-M0010-G020 CDX-M0010-G100
102990-354EA 192.46 USD
102990-354 102990-356
7-Acetoxy-4-methylcoumarin ≥99% (by NMR)
7-Acetoxy-4-methylcoumarin

Fluorogenic substrate for esterases, broadly applied to various types of esterases, including carboxyesterases. Measurement of intracellular pH in rat proximal convoluted tubule.

Formula: C₁₂H₁₀O₄
MW: 218.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006865
CAS Number: 2747-05-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 99% (NMR)
Appearance/Color White powder.
Solubility Soluble in DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR