(R)-(-)-Methyl-3-hydroxybutyrate

Supplier: ADIPOGEN CORP MS

Synonyms: (R)-Methyl 3-hydroxybutanoate, Methyl (R)-3-hydroxybutyrate, Methyl (R)-(-)-3-Hydroxybutyrate

CDX-H0102-G001 CDX-H0102-G005
102990-090EA 88.62 USD
102990-090 102990-092
(R)-(-)-Methyl-3-hydroxybutyrate
(R)-(-)-Methyl-3-hydroxybutyrate

Important optically active compound for the preparation of various bifunctional building blocks. Can be used in the synthesis of chiral ß-lactams. Can undergo aldol-type reactions with aldehydes with either syn- or anti-selectivity according to the conditions.

Formula: C₅H₁₀O₃
MDL Number: MFCD00063289
CAS Number: 3976-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR