Sodium-DL-3-hydroxybutyrate

Supplier: ADIPOGEN CORP MS

Synonyms: Sodium 3-hydroxybutanoate, DL-3-Hydroxybutyric acid Sodium salt

CDX-H0080-G001 CDX-H0080-G010
102990-266EA 54 USD
102990-266 102990-268
Sodium-DL-3-hydroxybutyrate
Sodium-DL-3-hydroxybutyrate

Neuroprotective compound in experimental models of neurodegenerative diseases. Shown to be oxidized as an energetic substrate in the brain during starvation. May inhibit brain glucose oxidation, which may account for the suppression of hepatic glucose production observed after administration of ketone bodies in humans.

Formula: C₄H₈O₃
MW: 126.09 g/mol
MDL Number: MFCD00016716
CAS Number: 150-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR