5-Hydroxynicotinic acid

Supplier: ADIPOGEN CORP MS

Synonyms: 5-Hydroxypyridine-3-carboxylic acid, 5-Hydroxy-3-pyridinecarboxylic acid

CDX-H0052-G001 CDX-H0052-G005 CDX-H0052-G025
102990-244EA 105.23 USD
102990-244 102990-246 102990-248
5-Hydroxynicotinic acid
5-Hydroxynicotinic acid

A nicotinic acid analog. Substrate analog MHPCO, used in the investigation of the hydroxylation mechanism of the flaovoprotein 2-methyl-3-hydroxypyridine-5-carboxylic acid oxygenase (MHPCO). Also used as building block for synthesis. Hydroxynicotinic acid is formed by the enzymatic action of nicotinate with nicotinic acid hydroxylase or nicotinate hydroxylase.

Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00129117
CAS Number: 27828-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR