α-D-Galacturonic acid monohydrate ≥98% (by titrimetric analysis)

Supplier: ADIPOGEN CORP MS

Synonyms: (2S,3R,4S,5R)-2,3,4,5-Tetrahydroxy-6-oxohexanoic acid hydrate, D-Galacturonic acid monohydrate

CDX-G0004-G025 CDX-G0004-G050 CDX-G0004-G250
102990-038EA 258.92 USD
102990-038 102990-040 102990-042
α-D-Galacturonic acid monohydrate ≥98% (by titrimetric analysis)
alpha-D-Galacturonic acid monohydrate

Highly purified product form pectine.

Formula: C₆H₁₀O₇·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00071585
CAS Number: 91510-62-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >98% (Titration)
Appearance/Color White crystalline powder.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR