4-Fluoro-2,1,3-benzoxadiazole

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Fluorobenzofurazan

CDX-F0213-G003 CDX-F0213-G010
102990-028EA 263.08 USD
102990-028 102990-030
4-Fluoro-2,1,3-benzoxadiazole
4-Fluoro-2,1,3-benzoxadiazole

4-Fluoro-2,1,3-benzoxadiazole is a fluorescent benzofurazan derivative.

Formula: C₆H₃FN₂O
MW: 138.10 g/mol
MDL Number: MFCD03425566
CAS Number: 29270-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR