D-Fructose-6-phosphate disodium salt ≥98%

Supplier: ADIPOGEN CORP MS
Danger

CDX-F0212-M500 CDX-F0212-G005 CDX-F0212-G025
102990-022EA 166.15 USD
102990-022 102990-024 102990-026
D-Fructose-6-phosphate disodium salt ≥98%
D-Fructose-6-phosphate disodium salt

A glycolytic intermediate formed by the isomerization of glucose-6-phosphate.

Formula: C₆H₁₁Na₂O₉P
MW: 304.098 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00135619
CAS Number: 26177-86-6

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color White solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR