4-Fluororesorcinol

Supplier: ADIPOGEN CORP MS

Synonyms: 1-Fluoro-2,4-dihydroxybenzene

CDX-F0047-M500 CDX-F0047-G001 CDX-F0047-G010
102989-968EA 155.08 USD
102989-968 102989-970 102989-972
4-Fluororesorcinol
4-Fluororesorcinol

Building block for synthesis.

Formula: C₆H₅FO₂
MW: 128.10 g/mol
MDL Number: MFCD03789074
CAS Number: 103068-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR