Fluorescein-5-maleimide ≥90% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 1-(3',6'-Dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-5-yl)-1H-pyrrole-2,5-dione, 1-(3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthen]-5-yl)-1H-pyrrole-2,5-dione, N-(5-Fluoresceinyl)-maleimide, 5-Maleimidofluorescein

CDX-F0004-M050 CDX-F0004-M500
102987-924EA 290.77 USD
102987-924 102987-926
Fluorescein-5-maleimide ≥90% (by HPLC)
Fluorescein-5-maleimide

Green fluorescent thiol-reactive dye (Ex/Em: 494/519nm) for bioconjugation, widely used as protein and peptide fluorescent labeling reagent, easily conjugating fluorescein. The reaction requires very mild condition and is highly specific.


The maleimide group reacts optimally with sulfhydryl groups on cysteine side chains at pH 7, forming a stable thioether bond. The compound can be used to label proteins for the detection of conformational changes, assembly of multisubunit complexes, and ligand-binding processes.

Formula: C₂₄H₁₃NO₇
MW: 427.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077345
CAS Number: 75350-46-8

Order Now

Specification Test Results

Source/Host Synthetic
Purity ≥90% (HPLC)
Identity Determined by 1H-NMR
Appearance Orange to brown solid
Solubility Soluble in DMF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR