Fluorescamine

Supplier: ADIPOGEN CORP MS

Synonyms: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione

CDX-F0003-M100 CDX-F0003-M250 CDX-F0003-G001
102987-918EA 74.77 USD
102987-918 102987-920 102987-922
Fluorescamine
Fluorescamine

A non-fluorescent reagent that reacts readily under mild conditions with primary amino groups in amino acids and peptides to form stable and highly fluorescent derivatives. Low background due to hydrolysis. Useful in fluorometric assays of amino acids, protein, and proteolytic enzymes. Effectively blocks newly generated amino termini in protein sequence analysis by the Beckman automated sequencer. Spectral data: Exc. 390nm/Em. 475 nm in 0.5 M borate buffer, pH 8.5, after derivatization with L-leucine.

Formula: C₁₇H₁₀O₄
MW: 278.26 g/mol
MDL Number: MFCD00005928
CAS Number: 38183-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR