7-Fluorobenzo-2-oxa-13-diazole-4-sulfonic acid ammonium salt

Supplier: ADIPOGEN CORP MS

Synonyms: Ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate

CDX-F0002-M005 CDX-F0002-M025 CDX-F0002-M250
102987-912EA 90 USD
102987-912 102987-914 102987-916
7-Fluorobenzo-2-oxa-13-diazole-4-sulfonic acid ammonium salt
7-Fluorobenzo-2-oxa-13-diazole-4-sulfonic acid ammonium salt

Fluorescent probe for thiols widely used in pre-column labeling of biological thiols for HPLC.

Formula: C₆H₆FN₃O₄S
MW: 235.189 g/mol
MDL Number: MFCD02682109
CAS Number: 84806-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR