Dimethyl (1-Diazo-2-oxopropyl)phosphonate
Supplier: ADIPOGEN CORP MS
CDX-D0501-G001
CDX-D0501-G005
102989-836EA
263.08
USD
102989-836
102989-838
Dimethyl (1-Diazo-2-oxopropyl)phosphonate
Dimethyl (1-diazo-2-oxopropyl)phosphonate
Used in the Seyferth-Gilbert homologation which is a chemical reaction of an aryl ketone or aldehyde with dimethyl- or diazomethyl-phosphonate and potassium tert-butoxide to give substituted alkynes.
Formula:
C₅H₉N₂O₄P MW: 192.11 g/mol |
MDL Number:
MFCD07368360 CAS Number: 90965-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...