((Difluoromethyl)sulfonyl)benzene ≥98%
Supplier: ADIPOGEN CORP MS
Synonyms:
Difluoromethyl phenyl sulphone
Building block for synthesis. Has been used for nucleophilic difluoro(phenylsulfonyl)methylation of carbonyls, reductive silylation and the preparation of trifluoro- and difluoromethylsilanes, fluoroalkylation/chloroalkylation of alpha,beta-enones, arynes, acetylenic ketones and other Michael acceptors and difluoromethylation of primary alkyl halides . It is also used in preparation of alpha-difluoromethyl amines, anti-difluoropropanediols, beta-difluoromethylated and beta-difluoromethylenated alcohols and amines, difluoroalkenes, difluoromethyl alcohol derivatives and fluoromethylated vicinal ethylenediamines.
Formula:
C₇H₆F₂O₂S MW: 192.18 g/mol |
MDL Number:
MFCD01050170 CAS Number: 1535-65-5 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Colorless to light yellow semi-solid or solid. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...