2,4-Difluororesorcinol

Supplier: ADIPOGEN CORP MS

Synonyms: 2,4-Difluorobenzene-1,3-diol

CDX-D0329-M500 CDX-D0329-G001 CDX-D0329-G005
102989-768EA 139.85 USD
102989-768 102989-770 102989-772
2,4-Difluororesorcinol
2,4-Difluororesorcinol

Fluorinated building block for dye synthesis. Key precusor for preparing Pacific Blue dyes. Precursor that can be used for preparing fluorescent dyes such as fluorescein, coumarin and resorufin analogs.

Formula: C₆H₄F₂O₂
CAS Number: 195136-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR