4-(Dimethylamino)cinnamaldehyde

Supplier: ADIPOGEN CORP MS

CDX-D0310-G005 CDX-D0310-G025
102989-750EA 105.23 USD
102989-750 102989-752
4-(Dimethylamino)cinnamaldehyde
4-(Dimethylamino)cinnamaldehyde

Primarily used as a histological dye to detect indoles. Reagent for the determination of aromatic amines. It is used for the rapid identification of bacteria containing apotryptophanase and tryptophanase enzyme systems. It is also particularly useful for localization of proanthocyanidins compounds in plants, resulting in a blue staining. Has been developed for flavanoids in beer and is used in forensics.

Formula: C₁₁H₁₃NO
MDL Number: MFCD00007002
CAS Number: 6203-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR