(Difluoromethyl)trimethylsilane

Supplier: ADIPOGEN CORP MS

CDX-D0301-G001 CDX-D0301-G002 CDX-D0301-G010
102989-736EA 245.08 USD
102989-736 102989-738 102989-740
(Difluoromethyl)trimethylsilane
(Difluoromethyl)trimethylsilane

Fluorination reagent.

Formula: C₄H₁₀F₂Si
MW: 124.21 g/mol
MDL Number: MFCD19381658
CAS Number: 65864-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR