1,7-Dimethyluric acid
Supplier: ADIPOGEN CORP MS
Synonyms:
7,9-Dihydro-1,7-dimethyl-1H-purine-2,6,8(3H)-trione
CDX-D0295-M050
CDX-D0295-M250
102989-728EA
166.15
USD
102989-728
102989-730
1,7-Dimethyluric acid
1,7-Dimethyluric acid
A metabolite of caffeine with antioxidant and neuroprotective activities. Does not inhibit the differentiation of 3T3-L1 pre-adipocytes to mature adipocytes, but suppress the intracellular lipid accumulation after complete differentiation in a dose-dependent manner.
Formula:
C₇H₈N₄O₃ MW: 196.17 g/mol |
CAS Number:
33868-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...