N-Succinimidyl 4-[4-(Dimethylamino)phenylazo]benzoate

Supplier: ADIPOGEN CORP MS

Synonyms: DABCYL acid, SE, DABCYL N-Succinimidyl Ester, 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 4-((4-(dimethylamino)phenyl)diazenyl)benzoate

CDX-D0236-M025 CDX-D0236-M100 CDX-D0236-G001
102989-688EA 91.38 USD
102989-688 102989-690 102989-692
N-Succinimidyl 4-[4-(Dimethylamino)phenylazo]benzoate
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate

N-terminal modification of peptides in automated synthesis.

Formula: C₁₉H₁₈N₄O₄
MW: 366.38 g/mol
MDL Number: MFCD00467415
CAS Number: 146998-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR