1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: DiI, DiI perchlorate

CDX-D0230-M100 CDX-D0230-G001
102989-684EA 210.46 USD
102989-684 102989-686
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate ≥98% (by TLC)
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate

Endoplasmic reticulum membrane stain. Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons; provides intense, long-lasting staining of live neurons in vivo and in vitro.

Formula: C₅₉H₉₇ClN₂O₄
MW: 933.885 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00142354
CAS Number: 41085-99-8

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (TLC)
Appearance/Color Burgundy to purple powder or crystalline powder.
Solubility Soluble in DMSO, DMF or methanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR