5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)

Supplier: ADIPOGEN CORP MS

Synonyms: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid

CDX-D0156-G001 CDX-D0156-G005 CDX-D0156-G025
102989-638EA 42.92 USD
102989-638 102989-640 102989-642
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)

A sensitive reagent for measuring the free sulfhydryl content in proteins, peptides, and tissues. Used to characterize reactive thiol groups and photometric determination of thiols and for measuring low-molecular mass thiols such as glutathione in both pure solutions and biological samples, such as blood. It can also measure the number of thiol groups on proteins. Through reaction with aliphatic thiol groups a mixed disulfide of protein thiol and one mole of 2-nitro-5-thiobenzoate per mole of protein sulfhydryl group is being formed. DTNB has little absorbance. Reaction with -SH groups on proteins (from any solvent accessible Cys) under mild alkaline conditions (pH 7-8) produces the 2-nitro-5-thiobenzoate anion, which gives an intense yellow color with an absorption maximum at 409.5nm (Extinction coefficient: 14150 M-1*cm-1). Sensitive to various buffer ions, therefore, the extinction coefficient used to calculate the number of sulfhydryl groups must be matched to the reaction conditions. In case the thiol groups are in disulfide bonds, they must be reduced under anaerobic conditions prior to reaction with DTNB.

Formula: C₁₄H₈N₂O₈S₂
MW: 396.36 g/mol
MDL Number: MFCD00007140
CAS Number: 69-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR