(S)-2,4-Diaminobutanoic acid dihydrochloride

Supplier: ADIPOGEN CORP MS

Synonyms: L-2,4-Diaminobutanoic acid dihydrochloride, H-Dab-OH 2HCl, L-2,4-Diaminobutyric acid dihydrochloride, (2S)-2,4-Diaminobutanoic acid dihydrochloride, H-Daba-OH·2HCl, (S)-(+)-2,4-Diaminobutyric acid dihydrochloride

CDX-D0154-G001 CDX-D0154-G005
102987-906EA 78.92 USD
102987-906 102987-908
(S)-2,4-Diaminobutanoic acid dihydrochloride
(S)-2,4-Diaminobutanoic acid dihydrochloride

Unnatural amino acid derivative. Pharmacological tool and potential chiral building block used as an internal standard for amino acid analysis. Suitable reagent used for the differentiation of beta-N-methylamino-L-alanine from the diamino acids by using HPLC-FD, UHPLC-UV, UHPLC-MS, and triple quadrupole tandem mass spectrometry (UHPLC-MS/MS). Used in the quantification of neurotoxin beta-N-methylamino-L-alanine (BMAA) in seafood. It has been found to inhibit GABA transaminase (ABAT), producing elevated levels of GABA and to have an antitumor activity.

Formula: C4H12Cl2N2O2
MW: 191.06 g/mol
MDL Number: MFCD01632031
CAS Number: 1883-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR