Dihydrofluorescein diacetate
Supplier: ADIPOGEN CORP MS
Synonyms:
2-(3,6-Diacetoxy-9H-xanthen-9-yl)benzoic acid
Dihydrofluorescein diacetate is a fluorescent probe for detecting intracellular oxidants. It is hydrolyzed by cellular esterases to dihydrofluorescein and is then oxidized to fluorescein primarily by H2O2. Dihydrofluorescein diacetate might be reactive toward a broad range of oxidizing reactions that may be increased during intracellular oxidant stress. Cell-loading studies indicate that dihydrofluorescein achieves higher intracellular concentrations than the other redox sensors such as Dihydrorhodamine 123 (Ex/Em: 490/514nm).
Formula:
C₂₄H₁₈O₇ MW: 418.401 g/mol |
MDL Number:
MFCD00005056 CAS Number: 35340-49-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...