7-(Diethylamino)coumarin-3-carboxylic Acid
Supplier: ADIPOGEN CORP MS
Synonyms:
7ACC1, 7-(Diethylamino)-2-oxo-2H-chromene-3-carboxylic acid, DEAC, acid
CDX-D0036-M100
CDX-D0036-M200
CDX-D0036-G001
102987-688EA
146.77
USD
102987-688
102987-690
102987-692
7-(Diethylamino)coumarin-3-carboxylic Acid
7-(Diethylamino)coumarin-3-carboxylic acid
7-(Diethylamino)-coumarin-3-carboxylic acid is a reagent for the derivatisation of amines and proteins. It has also been used for labelling synthetic peptides for high-throughput detection.
Formula:
C₁₄H₁₅NO₄ MW: 261.28 g/mol |
MDL Number:
MFCD00068721 CAS Number: 50995-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...