Disodium 4,4'-Diazidostilbene-2,2'-disulfonate Tetrahydrate

Supplier: ADIPOGEN CORP MS

Synonyms: 4,4'-Diazidostilbene-2,2'-disulphonic Acid Disodium Salt

CDX-D0022-G010 CDX-D0022-G050 CDX-D0022-G250
102987-654EA 88.62 USD
102987-654 102987-656 102987-658
Disodium 4,4'-Diazidostilbene-2,2'-disulfonate Tetrahydrate
Disodium-4,4'-diazidostilbene-2,2'-disulfonate tetrahydrate

A reagent for enzyme immobilisation on fibroin by photo-crosslinking. The reagent is water-soluble over a wide pH range.

Formula: C₁₄H₁₀N₆O₆S₂
MW: 466.35 g/mol
MDL Number: MFCD02091646
CAS Number: 2718-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR