(-)-[(8,8-Dichlorocamphoryl)-sulfonyl]-oxaziridine

Supplier: ADIPOGEN CORP MS

Synonyms: (4aR,7R,8aS)-8,8-Dichloro-9,9-dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide

CDX-D0021-G005 CDX-D0021-G025
102989-634EA 333.69 USD
102989-634 102989-636
(-)-[(8,8-Dichlorocamphoryl)-sulfonyl]-oxaziridine
(-)-[(8,8-Dichlorocamphoryl)sulfonyl]oxaziridine

Chiral oxidizing agent. Used for asymmetric synthesis as chiral catalyst. Hydroxylation reagent.

Formula: C₁₀H₁₃Cl₂NO₃S
MW: 298.178 g/mol
MDL Number: MFCD00243016
CAS Number: 139628-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR