3,3'-Dipropylthiacarbocyanine iodide

Supplier: ADIPOGEN CORP MS

CDX-D0007-M250 CDX-D0007-M500
102989-606EA 113.54 USD
102989-606 102989-608
3,3'-Dipropylthiacarbocyanine iodide
3,3'-Dipropylthiacarbocyanine iodide

Carbocyanine dyes, particularly thiacyanines such as DiSC3(3) can inhibit respiration and may therefore be relatively cytotoxic. 3,3'-dipropyl-thiadicarbocyanine iodide is sensitive to membrane potential, fluorescence response to depolarization depends on the staining concentration and detection method. Selected applications are: -Calcium channels and other ion transport systems -Mitochondrial activity -Neurons and brain tissue -Membrane potential in intact yeast cells

Formula: C₂₃H₂₅IN₂S₂
MW: 520.491 g/mol
CAS Number: 53336-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR