2',7'-Dichlorofluorescein

Supplier: ADIPOGEN CORP MS

Synonyms: 2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one

CDX-D0004-G010 CDX-D0004-G025 CDX-D0004-G100
102989-600EA 139.85 USD
102989-600 102989-602 102989-604
2',7'-Dichlorofluorescein
2',7'-Dichlorofluorescein

Fluorescent dye. Spectral data: Extinction 504nm/Emmission 529nm, in 0.1 M Tris pH 8.0. Used for the determination of carbohydrates by fluorescence densitometry after TLC, in the determination of H2O2 by chemiluminescence, and as a fluorescent additive used in indirect fluorimetric detection in OTC-HPLC. Used as a component/substrate of the 2′,7′-dichlorofluorescein diacetate (DCFH-DA) assay to quantitate reactive oxygen species (ROS).

Formula: C₂₀H₁₀Cl₂O₅
MW: 401.20 g/mol
MDL Number: MFCD00005047
CAS Number: 76-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR