4-(Chloromethyl)benzonitrile

Supplier: ADIPOGEN CORP MS

CDX-C0313-M500 CDX-C0313-G010
102989-596EA 42.92 USD
102989-596 102989-598
4-(Chloromethyl)benzonitrile
4-(Chloromethyl)benzonitrile

Building block for synthesis.

Formula: C₈H₆ClN
MW: 151.60 g/mol
MDL Number: MFCD00019761
CAS Number: 874-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR