2-(Chloromethyl)benzoic acid

Supplier: ADIPOGEN CORP MS

CDX-C0307-G001 CDX-C0307-G005 CDX-C0307-G025
102989-576EA 54 USD
102989-576 102989-578 102989-580
2-(Chloromethyl)benzoic acid
2-(Chloromethyl)benzoic acid

Building block for synthesis.

Formula: C₈H₇ClO₂
MDL Number: MFCD06208434
CAS Number: 85888-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR