2-Chloro-5-(chloromethyl)thiazole

Supplier: ADIPOGEN CORP MS

CDX-C0232-G001 CDX-C0232-G005
102989-556EA 139.85 USD
102989-556 102989-558
2-Chloro-5-(chloromethyl)thiazole
2-Chloro-5-(chloromethyl)thiazole

Intermediate in the preparation of agrochemicals/pesticides.

Formula: C₄H₃Cl₂NS
MDL Number: MFCD01073549
CAS Number: 105827-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR