2-Chlorotropone

Supplier: ADIPOGEN CORP MS

Synonyms: 2-Chlorocyclohepta-2,4,6-trienone, 2-Chloro-2,4,6-cycloheptatrien-1-one

CDX-C0181-G001 CDX-C0181-G002 CDX-C0181-G010
102989-530EA 139.85 USD
102989-530 102989-532 102989-534
2-Chlorotropone
2-Chlorotropone

Building block for synthesis.

Formula: C₇H₅ClO
MW: 140.57 g/mol
MDL Number: MFCD00182497
CAS Number: 3839-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR