(S)-(+)-Carvone

Supplier: ADIPOGEN CORP MS

Synonyms: D-p-Mentha-1(6),8-dien-2-one, D(+)-Carvone, (S)-2-Methyl-5-(prop-1-en-2-yl)cyclohex-2-enone, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-one

CDX-C0169-G005 CDX-C0169-G100 CDX-C0169-G500
102989-512EA 54 USD
102989-512 102989-514 102989-516
(S)-(+)-Carvone
(S)-(+)-Carvone

Terpene standard for GC.

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD00062997
CAS Number: 2244-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR