8-Chlorotheophylline

Supplier: ADIPOGEN CORP MS

CDX-C0153-G100 CDX-C0153-K001
102989-504EA 96.92 USD
102989-504 102989-506
8-Chlorotheophylline
8-Chlorotheophylline

Stimulant slightly ionotropic drug of the xanthine chemical class, with physiological effects similar to caffeine. It is combined with pharmaceutical drugs to form stable salts.

Formula: C₇H₇ClN₄O₂
MDL Number: MFCD00005581
CAS Number: 85-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR