1-Chloro-1,3-dihydro-3,3-dimethyl-1,2-benziodoxole

Supplier: ADIPOGEN CORP MS

CDX-C0136-M100 CDX-C0136-M500
102989-492EA 139.85 USD
102989-492 102989-494
1-Chloro-1,3-dihydro-3,3-dimethyl-1,2-benziodoxole
1-Chloro-1,3-dihydro-3,3-dimethyl-1,2-benziodoxole

A Hypervalent iodine reagent for oxidation reactions.

Formula: C₉H₁₀ClIO
MDL Number: MFCD21648251
CAS Number: 69352-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR