(5(6)-TAMRA

Supplier: ADIPOGEN CORP MS

Synonyms: TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt

CDX-C0038-M100 CDX-C0038-G002
102987-598EA 276.92 USD
102987-598 102987-600
(5(6)-TAMRA
5(6)-TAMRA

Mixed isomers of carboxytetramethylrhodamine free acid. Tetramethylrhodamine (TMR) is one of the fluorophores most often used for preparing peptide, protein, nucleotide and nucleic acid conjugates. TMR dyes have been widely used as acceptors for FAM fluorophores in a variety of FRET studies. The dye can be coupled to primary or secondary amines via standard peptide chemistry.

Formula: C₂₅H₂₂N₂O₅
MW: 430.45 g/mol
MDL Number: MFCD16661194
CAS Number: 98181-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR