6-Carboxyfluorescein

Supplier: ADIPOGEN CORP MS

CDX-C0016-M050 CDX-C0016-M200 CDX-C0016-G001
102987-570EA 123.23 USD
102987-570 102987-572 102987-574
6-Carboxyfluorescein
6-Carboxyfluorescein

The amine-reactive fluorescein derivatives are probably the most common fluorescent derivatization reagents for covalently labeling proteins. Carboxyfluorescein is better retained in cells than is fluorescein, its pKa of ~6.5 is lower than the cytosolic pH of most cells (pH ~6.8-7.4). Carboxyfluorescein is commonly employed as a polar tracer.

Formula: C₂₁H₁₂O₇
MDL Number: MFCD00036873
CAS Number: 3301-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR