5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)

Supplier: ADIPOGEN CORP MS

Synonyms: 2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate, 5-FAM-N-succinimidyl ester, 5-Carboxyfluorescein NHS ester, 5-FAM SE

CDX-C0015-M010 CDX-C0015-M025 CDX-C0015-M100
102987-556EA 271.38 USD
102987-556 102987-566 102987-568
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)

Popular green fluorescent cell-permeable dye. Covalently couples via its succinimidyl group to intracellular molecules. The amine-reactive succinimidyl ester of 5-FAM acid is a common fluorescent derivatization reagent for covalently labeling peptides, proteins and nucleotides. Due to this covalent coupling reaction it can be retained within cells for extremely long periods and due to this stable linkage, once incorporated within cells the dye is not transferred to adjacent cells. Yields carboxyamides which are more resistant to hydrolysis than fluorescein conjugates prepared with FITC. Better retained in cells than fluorescein. Used in fluorescent small molecule preparations. Spectral data: lambdaex 494 nm| lambdaem 520 nm (Buffer pH 9.0).

Formula: C₂₅H₁₅NO₉
MW: 473.39 g/mol
MDL Number: MFCD00077322
CAS Number: 92557-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR