5(6)-Carboxy-X-rhodamine
Supplier: ADIPOGEN CORP MS
Synonyms:
9-[2,4(or 2,5)-Dicarboxyphenyl]-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-xantheno[2,3,4-ij:5,6,7-i'j′]diquinolizin-18-ium inner salt, 5(6)-ROX
CDX-C0012-M025
CDX-C0012-M100
CDX-C0012-G001
102987-538EA
146.77
USD
102987-538
102987-540
102987-542
5(6)-Carboxy-X-rhodamine
5(6)-Carboxy-X-rhodamine
May be used as a fluorescent dye in nucleic acid analytical research applications.
Formula:
C₃₃H₃₀N₂O₅ MW: 534.612 g/mol |
MDL Number:
MFCD01862979 CAS Number: 198978-94-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...