4,4'-Bis(bromomethyl)biphenyl

Supplier: ADIPOGEN CORP MS

Synonyms: 4,4-Bis(bromomethyl)biphenyl

CDX-B0128-G001 CDX-B0128-G005 CDX-B0128-G025
102989-376EA 98.31 USD
102989-376 102989-378 102989-380
4,4'-Bis(bromomethyl)biphenyl
4,4'-Bis(bromomethyl)biphenyl

Building block for synthesis.

Formula: C₁₄H₁₂Br₂
MW: 340.06 g/mol
MDL Number: MFCD00094419
CAS Number: 20248-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR