4-tert-Butyl-1-(3-sulfopropyl)pyridinium hydroxide Inner salt hydrate

Supplier: ADIPOGEN CORP MS

Synonyms: 3-(4-tert.-Butyl-1-pyridinio)-1-propanesulphonate, 3-(4-(tert-Butyl)pyridin-1-ium-1-yl)propane-1-sulphonate, NDSB 256-4T

CDX-B0109-G001 CDX-B0109-G003
102989-368EA 108 USD
102989-368 102989-370
4-tert-Butyl-1-(3-sulfopropyl)pyridinium hydroxide Inner salt hydrate
4-tert-Butyl-1-(3-sulfopropyl)pyridinium hydroxide Inner salt hydrate

NDSB-256-4T is a non-detergent sulfobetaine (NDSB) that prevents protein aggregation and facilitates protein-folding, presumably by interacting with the early folding intermediates. Shown to improve the in vitro renaturation efficiency of reduced hen lysozyme and chemically unfolded tryptophan synthase beta2 subunit.

Formula: C₁₂H₁₉NO₃S
MDL Number: MFCD06411435
CAS Number: 570412-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR