2-Bromocycloheptanone
Supplier: ADIPOGEN CORP MS
CDX-B0101-M500
CDX-B0101-G001
CDX-B0101-G020
102989-362EA
108
USD
102989-362
102989-364
102989-366
2-Bromocycloheptanone
2-Bromocycloheptanone
Building block for synthesis.
Formula:
C₇H₁₁BrO |
MDL Number:
MFCD00015449 CAS Number: 766-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...