bisbenzimide H-33342 trihydrochloride (Hoechst 33342)

Supplier: ADIPOGEN CORP MS

Synonyms: Hoechst 33342, 2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, 2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole, bisBenzimide

CDX-B0030-M025 CDX-B0030-M500 CDX-B0030-G005
102989-326EA 40.15 USD
102989-326 102989-328 102989-330
bisbenzimide H-33342 trihydrochloride (Hoechst 33342)
bisbenzimide H-33342 trihydrochloride (Hoechst 33342)

Useful for staining DNA, chromosomes and nuclei. May be used for fluorescence microscopy or flow cytometry. Suitable for vital DNA staining of a variety of cell types and as a probe of membrane permeability in mammalian cells.

Formula: C₂₇H₂₈N₆O
MW: 561.93 g/mol
MDL Number: MFCD00012679
CAS Number: 23491-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR