3-(2-Benzothiazolyl)-7-(diethylamino)coumarin

Supplier: ADIPOGEN CORP MS

CDX-B0003-G001 CDX-B0003-G005 CDX-B0003-G010
102989-306EA 131.54 USD
102989-306 102989-308 102989-310
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin
3-(2-Benzothiazolyl)-7-(diethylamino)coumarin

Fluorescent dye which is widely used for staining polymer particles, but also organelles or materials used in medicine.

Formula: C₂₀H₁₈N₂O₂S
MW: 350.4341 g/mol
MDL Number: MFCD00041869
CAS Number: 38215-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR