Arsenazo III

Supplier: ADIPOGEN CORP MS

Synonyms: 2,7-Bis(2-arsonophenylazo)chromotropic acid, 2,7-Bis(2-arsonophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid, 2,2′-(1,8-Dihydroxy-3,6-disulphonaphthylene-2,7-bisazo)bisbenzenearsonic acid, Arsenazo Ⅲ

CDX-A0308-G001 CDX-A0308-G005 CDX-A0308-G010
102989-296EA 88.62 USD
102989-296 102989-298 102989-300
Arsenazo III
Arsenazo III

Arsenazo III is a calcium-sensitive dye. It is used for the determination of Ca2+ in micromolar amounts, to investigate calcium transport in permeabilized cells and determination of serum calcium. Also potentially useful for the spectrophotometric determination of uranium and thorium.

Formula: C₂₂H₁₈As₂N₄O₁₄S₂
MW: 776.38 g/mol
MDL Number: MFCD00036695
CAS Number: 1668-00-4
UN: 3465
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR