8-Amino-2-naphthol

Supplier: ADIPOGEN CORP MS

Synonyms: 1-Amino-7-naphthol

CDX-A0168-G025 CDX-A0168-G100
102989-222EA 139.85 USD
102989-222 102989-224
8-Amino-2-naphthol
8-Amino-2-naphthol

Chiral organo-catalyst. Chiral primary amine that can be used for the direct alpha-fluorination of alpha-branched aldehydes. Building block for synthesis.

Formula: C₁₀H₉NO
MW: 159.19 g/mol
MDL Number: MFCD00004031
CAS Number: 118-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR