Azomethine H sodium salt hydrate

Supplier: ADIPOGEN CORP MS

Synonyms: Sodium 5-hydroxy-4-((2-hydroxybenzylidene)amino)-7-sulphonaphthalene-2-sulphonate xhydrate, Azomethine-H monosodium salt hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate, Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate, Azomethine-H sodium hydrate

CDX-A0149-G005 CDX-A0149-G025 CDX-A0149-G100
102989-200EA 139.85 USD
102989-200 102989-202 102989-204
Azomethine H sodium salt hydrate
Azomethine H sodium salt hydrate

Probe for the colorimetric determination of boron in samples such us soils, plants, composts, manure, water, nutirent solution, glass or steel (microgram levels of boron). It forms an orange complex with boron in aqueous solution (absorption maxima at ~415nm). The detection range of boron in sample solutions is 1.0-6ppm. To detect boron in plant samples EDTA is used to mask copper, iron and aluminium ions. It is also used in electrocyclization reactions in the synthesis of martinellic acid, spirotryprostatin A and benzodiazepinones. Was shown to produce free radicals and might have anti-malarial and anti-cancer properties.

Formula: C₁₇H₁₂NNaO₈S₂·xH₂O
MW: 463.42 g/mol
MDL Number: MFCD00149588
CAS Number: 206752-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR