5-Aminofluorescein (isomer I) ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-A0018-M250 CDX-A0018-G001 CDX-A0018-G010
102989-120EA 56.77 USD
102989-120 102989-122 102989-124
5-Aminofluorescein (isomer I) ≥95% (by HPLC)
5-Aminofluorescein (isomer I)

The uronic acid residues of all known glycosaminoglycuronans react wit 5-aminofluorescein to yield fluorescent derivatives, which show fluorescence characteristics identical to those of 5-acetamidofluorescein (CDX-A0089). 5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020).

Formula: C₂₀H₁₃NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00005052
CAS Number: 3326-34-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC); 6-Isomer 1%
Appearance/Color Dark solid.
Solubility Soluble in acetone or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR