Fingolimod

Supplier: ADIPOGEN CORP MS

Synonyms: 2-Amino-2-(4-octylphenethyl)propane-1,3-diol hydrochloride, FTY-720 hydrochloride

AG-CR1-3587-M001 AG-CR1-3587-M005 AG-CR1-3587-M025
102988-664EA 13.85 USD
102988-664 102988-666 102988-668
Fingolimod
Fingolimod hydrochloride

Cell permeable immunosuppressor displaying lymphocyte sequestration properties and used for treatment in multiple sclerosis. Potent sphingosine 1-phosphate (S1P) receptor (S1P1, S1P3, S1P4, and S1P5) agonist when phosphorylated by sphingosine kinase. Sphingosine transporter Abcb1 and leukotriene C4 transporter Abcc1 activity enhancer. Cytosolic phospholipase A2 inhibitor, independent of sphingosine 1-phosphate receptors. Autophagy inducer. Apoptosis inducer.

Formula: C₁₉H₃₄ClNO₂
MW: 343.936 g/mol
MDL Number: MFCD00939512
CAS Number: 162359-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR