Resiquimod

Supplier: ADIPOGEN CORP MS

Synonyms: 1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, 1-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol, 4-Amino-2-(ethoxymethyl)-alpha,alpha-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol, R-848

AG-CR1-3582-M005 AG-CR1-3582-M025
102988-642EA 69.23 USD
102988-642 102988-644
Resiquimod
Resiquimod

Immune response modifier. Potent antitumor and antiviral compound. Stimulates antibody secretion and cytokine production. Used as adjuvant to increase the effectiveness of vaccines. Potent and selective synthetic ligand for Toll-like receptor 7 (TLR7) in mouse and for TLR7 and TLR8 in human. Activates immune cells via the TLR7/TLR8 MyD88-dependent signaling pathway and leads to the induction of NF-kappaB. NLRP3/NALP3 inflammasome activator, independent of TLRs and RIG-I. Upregulator of the opioid growth factor receptor. Used in the treatment of skin lesions such as herpes simplex virus.

Formula: C₁₇H₂₂N₄O₂
MW: 314.389 g/mol
MDL Number: MFCD00937759
CAS Number: 144875-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR