Nω-Methyl-L-arginine monoacetate ≥99%

Supplier: ADIPOGEN CORP MS

Synonyms: H-Arg(Me)-OH.AcOH, N5-(Methylamidino)-L-ornithine acetate salt, Nomega-Monomethyl-L-arginine acetate, L-NMMA, Ng-Methyl-L-Arginine Acetate Salt, Nomega-Methyl-L-arginine acetate, N-ω-Monomethyl-L-arginine monoacetate, NG-Monomethyl-L-arginine acetate salt

AG-CR1-3578-M005 AG-CR1-3578-M025 AG-CR1-3578-M100 AG-CR1-3578-G001
102988-620EA 34.62 USD
102988-620 102988-622 102988-624 102988-626
Nω-Methyl-L-arginine monoacetate ≥99%
Nω-Methyl-L-arginine monoacetate

Endothelium-derived relaxing factor inhibitor. Inhibits the generation of NO from arginine. Competitive, irreversible non-selective inhibitor of all three NOS I, II, and III isoforms (eNOS, iNOS and nNOS). Inhibits the synthesis of nitric oxide (NO) in a dose-dependent and enantiospecific fashion. Useful tool to study the role and the effects of NO in cardiovascular and gastrointestinal disorders, hypertension, diabetes, septic shock, inflammation, infection, stroke and neurodegenerative disorders.

Formula: C₇H₁₆N₄O₂·CH₃COOH
Storage Temperature: Freezer
MDL Number: MFCD00069311
CAS Number: 53308-83-1

Order Now

Specification Test Results

Purity 99% (TLC)
Appearance/Color White to off-white solid.
Solubility Soluble in water, ethanol, methanol or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR